missing translation for 'onlineSavingsMsg'
Learn More

Calcium Propionate 98.0+%, TCI America™

Catalog No. P050325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P050325G 25 g
P0503500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P050325G Supplier TCI America Supplier No. P050325G

Chemical Identifiers

CAS 4075-81-4
Molecular Formula C6H10CaO4
Molecular Weight (g/mol) 186.22
MDL Number MFCD00167354
InChI Key BCZXFFBUYPCTSJ-UHFFFAOYSA-L
Synonym calcium propionate, calcium dipropionate, calcium propanoate, mycoban, propanoic acid, calcium salt, bioban-c, caswell no. 151, propionic acid, calcium salt, unii-8ai80040kw, hsdb 907
PubChem CID 19999
ChEBI CHEBI:81716
IUPAC Name calcium dipropanoate
SMILES [Ca++].CCC([O-])=O.CCC([O-])=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 186.22
Percent Purity ≥98.0% (T)
Chemical Name or Material Calcium Propionate
EINECSNumber (9)-1227
RTECSNumber UE8450000
TSCA Yes