missing translation for 'onlineSavingsMsg'
Learn More

Calcium Propionate, Powder, FCC, 98-100.5%, Spectrum™ Chemical

CA166, 4075-81-4, C6H10CaO4

Supplier:  Spectrum Chemical Mfg Cor CA16625KG

 View more versions of this product

Catalog No. 18601363


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Calcium Propionate, Powder, FCC is used as a preservative in processed meats and baked goods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

4075-81-4
C6H10CaO4
2.5 kg
[Ca++].CCC([O-])=O.CCC([O-])=O
186.22
FCC
Poly Pail
100%
MFCD00167354
BCZXFFBUYPCTSJ-UHFFFAOYSA-L
calcium dipropanoate
98 to 100.5%
0.2%
5%

Chemical Identifiers

4075-81-4
186.22
BCZXFFBUYPCTSJ-UHFFFAOYSA-L
[Ca++].CCC([O-])=O.CCC([O-])=O
C6H10CaO4
MFCD00167354
calcium dipropanoate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions