missing translation for 'onlineSavingsMsg'
Learn More

Calcium Propionate, Powder, FCC, 98-100.5%, Spectrum™ Chemical

Catalog No. 18601362
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601362 500 g Poly Bottle
18601363 2.5 kg Poly Pail
18601364 12 kg Poly Pail
18601365 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601362 Supplier Spectrum Chemical Mfg Cor Supplier No. CA166500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

CA166, 4075-81-4, C6H10CaO4

Spectrum™ Chemical Calcium Propionate, Powder, FCC is used as a preservative in processed meats and baked goods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 4075-81-4
Molecular Formula C6H10CaO4
Molecular Weight (g/mol) 186.22
MDL Number MFCD00167354
InChI Key BCZXFFBUYPCTSJ-UHFFFAOYSA-L
IUPAC Name calcium dipropanoate
SMILES [Ca++].CCC([O-])=O.CCC([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 98 to 100.5%
Grade FCC
Insoluble Matter 0.2%
Packaging Poly Bottle
Water 5%