Learn More
Calcium propionate, 98+%
CAS: 4075-81-4 | C6H10CaO4 | 186.22 g/mol
Supplier: Thermo Scientific Chemicals 403900050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Calcium propionate | |
| >300.0°C | |
| 6.9 to 9.0 (10% soln.) | |
| Authentic | |
| Plastic drum | |
| (CH3CH2COO)2Ca | |
| 5 kg | |
| calcium propionate, calcium dipropionate, calcium propanoate, mycoban, propanoic acid, calcium salt, bioban-c, caswell no. 151, propionic acid, calcium salt, unii-8ai80040kw, hsdb 907 | |
| BCZXFFBUYPCTSJ-UHFFFAOYSA-L | |
| calcium;propanoate | |
| 19999 | |
| 186.22 | |
| 0.3% max. (in water) |
| 4075-81-4 | |
| White | |
| 4% max. (105°C, 2 Hrs) | |
| 98% min. (on dry substance) (Complexometry) | |
| C6H10CaO4 | |
| MFCD00167354 | |
| 15, 1700 | |
| Solubility in water: 260g/L (20°C). Other solubilities: slightly soluble in methanol and ethanol,practically insoluble in acetone and benzene | |
| [Ca++].CCC([O-])=O.CCC([O-])=O | |
| 186.22 | |
| CHEBI:81716 | |
| 98+% | |
| Crystalline Powder or Crystals |
Chemical Identifiers
| 4075-81-4 | |
| 186.22 | |
| BCZXFFBUYPCTSJ-UHFFFAOYSA-L | |
| 19999 | |
| calcium;propanoate |
| C6H10CaO4 | |
| MFCD00167354 | |
| calcium propionate, calcium dipropionate, calcium propanoate, mycoban, propanoic acid, calcium salt, bioban-c, caswell no. 151, propionic acid, calcium salt, unii-8ai80040kw, hsdb 907 | |
| CHEBI:81716 | |
| [Ca++].CCC([O-])=O.CCC([O-])=O |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 223-795-8
RUO – Research Use Only