missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Phthalate Hydrate 98.0+%, TCI America™
$190.13 - $190.13
Chemical Identifiers
| CAS | 5793-85-1 |
|---|---|
| Molecular Formula | C8H4CaO4 |
| Molecular Weight (g/mol) | 204.19 |
| MDL Number | MFCD08276328 |
| InChI Key | PYSZASIZWHHPHJ-UHFFFAOYSA-L |
| Synonym | Phthalic Acid Calcium Salt |
| PubChem CID | 91659099 |
| IUPAC Name | calcium benzene-1,2-dicarboxylate |
| SMILES | [Ca++].[O-]C(=O)C1=CC=CC=C1C([O-])=O |
Chemical Identifiers
| 5793-85-1 | |
| 204.19 | |
| PYSZASIZWHHPHJ-UHFFFAOYSA-L | |
| 91659099 | |
| [Ca++].[O-]C(=O)C1=CC=CC=C1C([O-])=O |
| C8H4CaO4 | |
| MFCD08276328 | |
| Phthalic Acid Calcium Salt | |
| calcium benzene-1,2-dicarboxylate |
Specifications
| 5793-85-1 | |
| C8H4CaO4 | |
| 5 g | |
| PYSZASIZWHHPHJ-UHFFFAOYSA-L | |
| calcium benzene-1,2-dicarboxylate | |
| 91659099 | |
| ≥98.0% (T) | |
| Calcium Phthalate Hydrate |
| White | |
| MFCD08276328 | |
| Phthalic Acid Calcium Salt | |
| [Ca++].[O-]C(=O)C1=CC=CC=C1C([O-])=O | |
| 204.19 | |
| 204.19 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1272
TSCA : No