Learn More
Calcium Phosphate, Monobasic, Monohydrate, Powder, FCC, 15.9-17.7%, Spectrum™ Chemical
CA158, 10031-30-8, Ca(H2PO4)2·H2O
Supplier:  Spectrum Chemical Mfg Cor CA15812KG
 View more versions of this product
Catalog No. 18601356
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Calcium Phosphate, Monobasic, Monohydrate, Powder, FCC is used in the food industry as a leavening agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
10031-30-8
CaH2O9P2
ZBZJARSYCHAEND-UHFFFAOYSA-H
calcium hydrate diphosphate
15.9 to 17.7%
1%
100%
12 kg
O.[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
248.04
FCC
Poly Pail
Chemical Identifiers
10031-30-8
248.04
calcium hydrate diphosphate
CaH2O9P2
ZBZJARSYCHAEND-UHFFFAOYSA-H
O.[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Product Suggestions
Product Suggestions