missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phosphate, Monobasic, Monohydrate, Powder, FCC, 15.9-17.7%, Spectrum™ Chemical

Catalog No. p-7157237
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601353 125 g Poly Bottle
18601354 500 g Poly Bottle
18601355 2.5 kg Poly Pail
18601356 12 kg Poly Pail
18601357 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601353 Supplier Spectrum Chemical Mfg Cor Supplier No. CA158125GM
Only null left

CA158, 10031-30-8, Ca(H2PO4)2·H2O

Spectrum™ Chemical Calcium Phosphate, Monobasic, Monohydrate, Powder, FCC is used in the food industry as a leavening agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10031-30-8
Molecular Formula CaH2O9P2
Molecular Weight (g/mol) 248.04
InChI Key ZBZJARSYCHAEND-UHFFFAOYSA-H
IUPAC Name calcium hydrate diphosphate
SMILES O.[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

Specifications

CAS Max % 100%
Quantity 125 g
Percent Purity 15.9 to 17.7%
Grade FCC
Loss on Drying 1%
Packaging Poly Bottle