missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Phosphate, Monobasic, Monohydrate, Powder, FCC, 15.9-17.7%, Spectrum™ Chemical
CA158, 10031-30-8, Ca(H2PO4)2·H2O
Supplier: Spectrum Chemical Mfg Cor CA15825KG
Description
Spectrum™ Chemical Calcium Phosphate, Monobasic, Monohydrate, Powder, FCC is used in the food industry as a leavening agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 10031-30-8 | |
| CaH2O9P2 | |
| ZBZJARSYCHAEND-UHFFFAOYSA-H | |
| calcium hydrate diphosphate | |
| 15.9 to 17.7% | |
| 1% |
| 100% | |
| 2.5 kg | |
| O.[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O | |
| 248.04 | |
| FCC | |
| Poly Pail |
Chemical Identifiers
| 10031-30-8 | |
| 248.04 | |
| calcium hydrate diphosphate |
| CaH2O9P2 | |
| ZBZJARSYCHAEND-UHFFFAOYSA-H | |
| O.[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O |