missing translation for 'onlineSavingsMsg'
Learn More

Calcium perchlorate hydrate, Reagent Grade, Thermo Scientific Chemicals

$122.33 - $122.33

Chemical Identifiers

CAS 13477-36-6
Molecular Formula CaCl2O8
Molecular Weight (g/mol) 238.97
MDL Number MFCD00015971
InChI Key ZQAOXERLHGRFIC-UHFFFAOYSA-L
Synonym calcium perchlorate, calcium diperchlorate, perchloric acid,calcium salt, tetrahydrate 8ci,9ci, hydrated, perchloric acid, calcium salt, perchloric acid, calcium salt 2:1, ca.2clo4, calcium 2+ diperchlorate ion, calcium perchlorate hydrate 250g, perchloric acid calcium salt 2:1e
PubChem CID 61629
IUPAC Name calcium;diperchlorate
SMILES [Ca++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1165530
View Documents
Thermo Scientific Chemicals
01165530
250 g
Each for $122.33
Only null left
 
Description

Description

Calcium perchlorate hydrate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13477-36-6
238.97
ZQAOXERLHGRFIC-UHFFFAOYSA-L
61629
[Ca++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
CaCl2O8
MFCD00015971
calcium perchlorate, calcium diperchlorate, perchloric acid,calcium salt, tetrahydrate 8ci,9ci, hydrated, perchloric acid, calcium salt, perchloric acid, calcium salt 2:1, ca.2clo4, calcium 2+ diperchlorate ion, calcium perchlorate hydrate 250g, perchloric acid calcium salt 2:1e
calcium;diperchlorate

Specifications

13477-36-6
Crystalline
CaCl2O8
UN1455
Soluble in water.
[Ca++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
238.97
238.98 (Anhydrous)
Odorless
Calcium perchlorate hydrate
White
250 g
MFCD00015971
calcium perchlorate, calcium diperchlorate, perchloric acid,calcium salt, tetrahydrate 8ci,9ci, hydrated, perchloric acid, calcium salt, perchloric acid, calcium salt 2:1, ca.2clo4, calcium 2+ diperchlorate ion, calcium perchlorate hydrate 250g, perchloric acid calcium salt 2:1e
ZQAOXERLHGRFIC-UHFFFAOYSA-L
calcium;diperchlorate
61629
Reagent
Hygroscopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: CALCIUM PERCHLORATE

EINECSNumber : 236-768-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only