missing translation for 'onlineSavingsMsg'
Learn More

Medisca Calcium Pantothenate (D), USP (Dietary Supplement Grade)

Catalog No. 14186112
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186112 1 kg
14185510 25 g
14185513 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14186112 Supplier Medisca Supplier No. 087609
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Calcium Pantothenate (D), USP (Dietary Supplement Grade) is an odorless, slightly hygroscopic, white powder.

Calcium Pantothenate (D), USP is an odorless, slightly hygroscopic, white powder. It is freely soluble in water, soluble in glycerin and practically insoluble in alcohol.

Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.

Alternative name(s): Vitamin B5, Pantothenic Acid Calcium Salt

Specifications

Chemical Name or Material Calcium Pantothenate (D), USP (Dietary Supplement Grade)
Packaging Jar, 1500mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 137-08-6
Type Drug Standard
Molecular Weight (g/mol) 476.536
Quantity 1 kg
Percent Purity 98 to 102 % (Calcium Pantothenate)
Grade USP