missing translation for 'onlineSavingsMsg'
Learn More

Calcium oxalate monohydrate, 99%

Catalog No. p-3256942
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3243022 100 g
AA3243036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3243022 Supplier Thermo Scientific Chemicals Supplier No. 03243022
Only null left

CAS: 5794-28-5 | C2H2CaO5 | 146.11 g/mol

Calcium oxalate monohydrate is used as a precursor in the production of oxalic acid, organic oxalates, ceramic glazes, and for rare-earth-metal separation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5794-28-5
Molecular Formula C2H2CaO5
Molecular Weight (g/mol) 146.11
MDL Number MFCD00150039
InChI Key LQHWSGSWNOHVHO-UHFFFAOYSA-L
Synonym calcium oxalate monohydrate, unii-4pp86kk527, calcium hydrate oxalate, calcium ethanedioate hydrate 1:1:1, acmc-1bmv0, ethanedioic acid, calcium salt 1:1 , monohydrate, calcium oxalate monohydrate 100g, ethanedioic acid,calcium salt, hydrate 1:?:?
PubChem CID 165350
IUPAC Name calcium;oxalate;hydrate
SMILES O.[Ca++].[O-]C(=O)C([O-])=O

Specifications

Melting Point 200°C
Physical Form Lump/Powder
Quantity 100 g
Assay Percent Range 99%
Solubility Information Slightly soluble in water.
Formula Weight 146.11 (128.10 Anhydrous)
Sensitivity Hygroscopic
Density 2.12
Chemical Name or Material Calcium oxalate, monohydrate
Hazard Category H302+H312
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
EINECSNumber 209-260-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only