missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium oxalate monohydrate, 98%, extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 200.0°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 500 g |
| Assay Percent Range | 98% |
| Linear Formula | (COO)2Ca·H2O |
| UN Number | 2811 |
| Merck Index | 14, 1685 |
| Solubility Information | Solubility in water: practically insoluble. Other solubilities: practically insoluble in acetic acid,soluble in dilute hcl or hno3 |
| Formula Weight | 146.11 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Harmful in contact with skin. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. |
RUO – Research Use Only