missing translation for 'onlineSavingsMsg'
Learn More

Calcium oxalate monohydrate, 98%, extra pure

Catalog No. AC403880050
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC403880050 5 g Glass bottle
AC403885000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC403880050 Supplier Thermo Scientific Chemicals Supplier No. 403880050
Only null left

CAS: 5794-28-5 | C2H2CaO5 | 146.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5794-28-5
Molecular Formula C2H2CaO5
Molecular Weight (g/mol) 146.11
MDL Number MFCD00150039
InChI Key LQHWSGSWNOHVHO-UHFFFAOYSA-L
Synonym calcium oxalate monohydrate, unii-4pp86kk527, calcium hydrate oxalate, calcium ethanedioate hydrate 1:1:1, acmc-1bmv0, ethanedioic acid, calcium salt 1:1 , monohydrate, calcium oxalate monohydrate 100g, ethanedioic acid,calcium salt, hydrate 1:?:?
PubChem CID 165350
IUPAC Name calcium;oxalate;hydrate
SMILES O.[Ca++].[O-]C(=O)C([O-])=O

Specifications

Melting Point 200.0°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 98%
Linear Formula (COO)2Ca·H2O
UN Number 2811
Merck Index 14, 1685
Solubility Information Solubility in water: practically insoluble. Other solubilities: practically insoluble in acetic acid,soluble in dilute hcl or hno3
Formula Weight 146.11
Percent Purity 98%
Grade Extra Pure
Loss on Drying 13% max. (200°C)
Packaging Glass bottle
Chemical Name or Material Calcium oxalate monohydrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only