missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Nitrate Tetrahydrate, ACS Reagent, Reagents
Calcium Nitrate Tetrahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C116550050F4
Specifications
| 132°C | |
| 561°C | |
| 13477-34-4 | |
| 50 kg | |
| Nitric Acid calcium salt | |
| O.[Ca++].[O-][N+]([O-])=O | |
| 236.15 | |
| Fiber Drum |
| Calcium Nitrate Tetrahydrate, ACS Reagent | |
| Colorless/White | |
| Solid | |
| CaH8N2O10 | |
| MWGCGFYACDTFSB-UHFFFAOYSA-N | |
| calcium bis(nitric acid) tetrahydrate | |
| ACS | |
| 1.82 |
Chemical Identifiers
| 13477-34-4 | |
| 236.15 | |
| Nitric Acid calcium salt | |
| O.[Ca++].[O-][N+]([O-])=O |
| CaH8N2O10 | |
| MWGCGFYACDTFSB-UHFFFAOYSA-N | |
| calcium bis(nitric acid) tetrahydrate |