missing translation for 'onlineSavingsMsg'
Learn More

Calcium Nitrate Tetrahydrate, ACS Reagent, Reagents

Catalog No. p-200020987
Change view
Click to view available options
Quantity:
125 g
500 g
50 lb.
50 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338060 125 g
01338059 500 g
01338058 50 kg
01338057 50 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01338060 Supplier Labchem Inc Supplier No. C1165500125D2
Only null left

Calcium Nitrate Tetrahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
Synonym Nitric Acid calcium salt
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O

Specifications

Boiling Point 132°C
Melting Point 561°C
Color Colorless/White
Physical Form Solid
Quantity 125 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.82
Chemical Name or Material Calcium Nitrate Tetrahydrate, ACS Reagent