missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Mesoxalate Trihydrate 98.0+%, TCI America™
Supplier: TCI America O00765G
Specifications
| Calcium Mesoxalate Trihydrate | |
| White | |
| MFCD00150291 | |
| Mesoxalic Acid Calcium Salt, Calcium Ketomalonate | |
| C(=O)(C(=O)O)C(=O)O.C(=O)(C(=O)[O-])C(=O)[O-].O.O.[Ca+2] | |
| 310.18 | |
| 174.12 | |
| Crystalline Powder |
| 21085-60-9 | |
| C6H6CaO12 | |
| 5 g | |
| QHOHAGRGHNXSFU-UHFFFAOYSA-L | |
| calcium;2-oxopropanedioate;2-oxopropanedioic acid;dihydrate | |
| 131674760 | |
| ≥98.0% (T) |
Chemical Identifiers
| 21085-60-9 | |
| 310.18 | |
| QHOHAGRGHNXSFU-UHFFFAOYSA-L | |
| 131674760 | |
| C(=O)(C(=O)O)C(=O)O.C(=O)(C(=O)[O-])C(=O)[O-].O.O.[Ca+2] |
| C6H6CaO12 | |
| MFCD00150291 | |
| Mesoxalic Acid Calcium Salt, Calcium Ketomalonate | |
| calcium;2-oxopropanedioate;2-oxopropanedioic acid;dihydrate |
Safety and Handling
EINECSNumber : (2)-0922&(2)-2755
TSCA : No