accessibility menu, dialog, popup

UserName

CAS RN 21085-60-9

CaOHOOOHO

CAS RN 21085-60-9

IUPAC Name: calcium 2-oxopropanedioic acid
Synonyms: calcium mesoxalic acid
Molecular Weight (g/mol): 158.12
Molecular Formula: C3H2CaO5
InChi Key: YYJSBERANQTJJH-UHFFFAOYSA-N
SMILES: [Ca++].OC(=O)C(=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results