missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium meso-Tartrate Trihydrate 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 25 g |
| Formula Weight | 188.15 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Calcium meso-Tartrate Trihydrate |
Safety and Handling
| EINECSNumber | (2)-1457 |
| TSCA | Yes |