CAS RN 815-79-2
IUPAC Name:
2,3-dihydroxybutanedioic acid calcium
Synonyms:
(.+-.)-tartaric acid calcium
Molecular Weight (g/mol):
190.16
Molecular Formula:
C4H6CaO6
InChi Key:
XTHSXAWAKVEMHZ-UHFFFAOYNA-N
SMILES:
[Ca].OC(C(O)C(O)=O)C(O)=O
1
–
1
of
1
results
1
Calcium meso-Tartrate Trihydrate 98.0+%, TCI America™
CAS: 815-79-2 Molecular Formula: C4H6CaO6 Molecular Weight (g/mol): 190.164 MDL Number: MFCD00151483 InChI Key: XTHSXAWAKVEMHZ-NUGIMEKKSA-N Synonym: Calcium Mesotartrate, Mesotartaric Acid Calcium Salt, meso-Tartaric Acid Calcium Salt PubChem CID: 131885529 IUPAC Name: calcium;(2R,3S)-2,3-dihydroxybutanedioic acid SMILES: C(C(C(=O)O)O)(C(=O)O)O.[Ca]