missing translation for 'onlineSavingsMsg'
Learn More

Calcium meso-Tartrate Trihydrate 98.0+%, TCI America™

Catalog No. T00041G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T00041G 1 g
T000425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T00041G Supplier TCI America Supplier No. T00041G
Only null left

Chemical Identifiers

CAS 815-79-2
Molecular Formula C4H6CaO6
Molecular Weight (g/mol) 190.164
MDL Number MFCD00151483
InChI Key XTHSXAWAKVEMHZ-NUGIMEKKSA-N
Synonym Calcium Mesotartrate, Mesotartaric Acid Calcium Salt, meso-Tartaric Acid Calcium Salt
PubChem CID 131885529
IUPAC Name calcium;(2R,3S)-2,3-dihydroxybutanedioic acid
SMILES C(C(C(=O)O)O)(C(=O)O)O.[Ca]

Specifications

Color White
Quantity 1 g
Formula Weight 188.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium meso-Tartrate Trihydrate
EINECSNumber (2)-1457
TSCA Yes