missing translation for 'onlineSavingsMsg'
Learn More

Calcium L-threonate, 98%

Catalog No. AAJ6627922
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6627922 100 g
AAJ6627936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6627922 Supplier Thermo Scientific Chemicals Supplier No. J6627922

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70753-61-6 | C8H14CaO10 | 310.27 g/mol

Calcium L-threonate is reactant involved in synthesis of cyclic hydroxamates and substituted oxazinanones and oxidative degradation to yield oxalate complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium L-threonate is reactant involved in synthesis of cyclic hydroxamates and substituted oxazinanones and oxidative degradation to yield oxalate complexes.

Solubility
Soluble in water. Sparingly soluble in methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 70753-61-6
Molecular Formula C8H14CaO10
Molecular Weight (g/mol) 310.27
MDL Number MFCD00077882,MFCD00150824
InChI Key ZJXGOFZGZFVRHK-UHFFFAOYNA-L
Synonym calcium l-threonate, l-threonic acid calcium salt, calcium 2r,3s-2,3,4-trihydroxybutanoate, calcium threonate, unii-hbb4ypo93u, hbb4ypo93u, calcium bis l-threonate, threonine, calcium salt, bis l-threonic acid calcium salt, butanoic acid, 2,3,4-trihydroxy-, calcium salt 2:1 , 2r,3s
PubChem CID 13388558
IUPAC Name calcium;(2R,3S)-2,3,4-trihydroxybutanoate
SMILES [Ca++].OCC(O)C(O)C([O-])=O.OCC(O)C(O)C([O-])=O

Specifications

Melting Point >300°C
Color Brown
Quantity 100 g
Solubility Information Soluble in water. Sparingly soluble in methanol.
Formula Weight 310.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Calcium L-threonate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only