missing translation for 'onlineSavingsMsg'
Learn More

Calcium iodide, ultra dry, 99.995% (metals basis)

CAS: 10102-68-8 | CaI2 | 293.887 g/mol

$347.77 - $1022.11

Chemical Identifiers

CAS 10102-68-8
Molecular Formula CaI2
Molecular Weight (g/mol) 293.887
MDL Number MFCD00010910
InChI Key UNMYWSMUMWPJLR-UHFFFAOYSA-L
Synonym calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g
PubChem CID 5462722
ChEBI CHEBI:81718
IUPAC Name calcium;diiodide
SMILES [Ca+2].[I-].[I-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4712114
View Documents
Thermo Scientific Chemicals
04712114
25 g
Each for $347.77
Only null left
 
AA4712122
View Documents
Thermo Scientific Chemicals
04712122
100 g
Each for $1,022.11
Only null left
 
Description

Description

It is used in photography. It is also used in cat food as a source of iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

10102-68-8
293.887
UNMYWSMUMWPJLR-UHFFFAOYSA-L
5462722
calcium;diiodide
CaI2
MFCD00010910
calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g
CHEBI:81718
[Ca+2].[I-].[I-]

Specifications

1100°C
10102-68-8
Beads
CaI2
14,1677
Soluble in water (64.6g/100 mL (0°C)). Soluble in acetone and alcohols.
[Ca+2].[I-].[I-]
293.887
CHEBI:81718
99.995%
Ampule
3.956 g/mL
(metals basis)
784°C
White to Yellow
25 g
MFCD00010910
calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g
UNMYWSMUMWPJLR-UHFFFAOYSA-L
calcium;diiodide
5462722
293.89
Odorless
Hygroscopic; Light sensitive
Calcium iodide, Ultra dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 233-276-8

TSCA : Yes

storageNote1 :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only