Learn More
Calcium iodate, pure
Oxidation reagent | CAS: 7789-80-2 | CaI2O6 | 389.89 g/mol
$229.23 - $229.23
Chemical Identifiers
| CAS | 7789-80-2 |
|---|---|
| Molecular Formula | CaI2O6 |
| Molecular Weight (g/mol) | 389.89 |
| InChI Key | UHWJJLGTKIWIJO-UHFFFAOYSA-L |
| Synonym | calcium iodate, lautarite, calcium diiodate, unii-l8mn4y57br, l8mn4y57br, iodic acid hio3 , calcium salt 2:1, autarite, iodic acid, calcium salt, calcium iodate ca io3 2, hsdb 986 |
| PubChem CID | 24619 |
| IUPAC Name | calcium;diiodate |
| SMILES | [O-]I(=O)=O.[O-]I(=O)=O.[Ca+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC318415000
|
Thermo Scientific Chemicals
318415000 |
500 g |
Each for $229.23
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7789-80-2 | |
| 389.89 | |
| calcium iodate, lautarite, calcium diiodate, unii-l8mn4y57br, l8mn4y57br, iodic acid hio3 , calcium salt 2:1, autarite, iodic acid, calcium salt, calcium iodate ca io3 2, hsdb 986 | |
| calcium;diiodate |
| CaI2O6 | |
| UHWJJLGTKIWIJO-UHFFFAOYSA-L | |
| 24619 | |
| [O-]I(=O)=O.[O-]I(=O)=O.[Ca+2] |
Specifications
| 540.0°C | |
| 500 g | |
| Ca(IO3)2 | |
| calcium iodate, lautarite, calcium diiodate, unii-l8mn4y57br, l8mn4y57br, iodic acid hio3 , calcium salt 2:1, autarite, iodic acid, calcium salt, calcium iodate ca io3 2, hsdb 986 | |
| UHWJJLGTKIWIJO-UHFFFAOYSA-L | |
| calcium;diiodate | |
| 24619 | |
| ≥96% (Iodometry) | |
| Calcium iodate |
| 7789-80-2 | |
| CaI2O6 | |
| 15,1678 | |
| Solubility in water: 2g/L (15°C). | |
| [O-]I(=O)=O.[O-]I(=O)=O.[Ca+2] | |
| 389.89 | |
| 389.89 | |
| Pure |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
EINECSNumber : 232-191-3
RUO – Research Use Only