missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium(II) Bis(nonafluorobutanesulfonyl)imide 95.0+%, TCI America™
$169.63 - $505.16
Chemical Identifiers
| CAS | 689282-13-1 |
|---|---|
| Molecular Formula | C16CaF36N2O8S4 |
| Molecular Weight (g/mol) | 1200.443 |
| InChI Key | WGPFQGXTGKSRPV-UHFFFAOYSA-N |
| Synonym | Bis(nonafluorobutanesulfonyl)imide Calcium(II) Salt |
| PubChem CID | 134128889 |
| IUPAC Name | calcium;bis(1,1,2,2,3,3,4,4,4-nonafluorobutylsulfonyl)azanide |
| SMILES | C(C(C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(F)(F)F)(F)F.C(C(C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(F)(F)F)(F)F.[Ca+2] |
Chemical Identifiers
| 689282-13-1 | |
| 1200.443 | |
| Bis(nonafluorobutanesulfonyl)imide Calcium(II) Salt | |
| calcium;bis(1,1,2,2,3,3,4,4,4-nonafluorobutylsulfonyl)azanide |
| C16CaF36N2O8S4 | |
| WGPFQGXTGKSRPV-UHFFFAOYSA-N | |
| 134128889 | |
| C(C(C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(F)(F)F)(F)F.C(C(C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(F)(F)F)(F)F.[Ca+2] |
Specifications
| 689282-13-1 | |
| C16CaF36N2O8S4 | |
| 1759 | |
| WGPFQGXTGKSRPV-UHFFFAOYSA-N | |
| calcium;bis(1,1,2,2,3,3,4,4,4-nonafluorobutylsulfonyl)azanide | |
| 134128889 | |
| ≥95.0% (T) | |
| Calcium(II) Bis(nonafluorobutanesulfonyl)imide |
| White | |
| 1 g | |
| Bis(nonafluorobutanesulfonyl)imide Calcium(II) Salt | |
| C(C(C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(F)(F)F)(F)F.C(C(C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(F)(F)F)(F)F.[Ca+2] | |
| 1200.443 | |
| 1200.44 | |
| Crystalline Powder |