accessibility menu, dialog, popup

UserName

CAS RN 689282-13-1

FFSOONSOOFFFFFFFFFFFFFFFFFFSOONSOOFFFFFFFFFFFFFFFFCa

CAS RN 689282-13-1

IUPAC Name: calcium bis(1,1,1,2,2,3,3,4,4-nonafluoro-4-{[(1,1,2,2,3,3,4,4,4-nonafluorobutanesulfonyl)azanidyl]sulfonyl}butane)
Synonyms: calcium bis(1,1,1,2,2,3,3,4,4-nonafluoro-4-[(1,1,2,2,3,3,4,4,4-nonafluorobutanesulfonyl)azanidylsulfonyl]butane)
Molecular Weight (g/mol): 1200.44
Molecular Formula: C16CaF36N2O8S4
InChi Key: WGPFQGXTGKSRPV-UHFFFAOYSA-N
SMILES: [Ca++].FC(F)(F)C(F)(F)C(F)(F)C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F.FC(F)(F)C(F)(F)C(F)(F)C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results