missing translation for 'onlineSavingsMsg'
Learn More

Calcium ICP-MS Standard, 10,000 ppm Ca in 5% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company MSCA10KN100

Catalog No. MSCA10KN-100


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Calcium ICP-MS Standard
471-34-1,7697-37-2,7732-18-5
85.0,10.52,2.13
1 mL = 10 mg Ca
CCaO3
Colorless
Approximately 100°C
Miscible with water
ICP Single-element Standard Solution
[Ca++].[O-]C([O-])=O
100.09
10112
Laboratory
Natural LDPE Bottle
10,000ppm Ca
89.0,10.95,2.22
Liquid
MFCD00010906
< 2
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Calibration Standard
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
100 mL
CHEBI:3311

Chemical Identifiers

471-34-1
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
CHEBI:3311
[Ca++].[O-]C([O-])=O
CCaO3
MFCD00010906
10112
calcium carbonate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions