Learn More
Calcium hypochlorite, tech., granular, 65% available chlorine
CAS: 7778-54-3 | CaCl2O2 | 142.98 g/mol
$71.93 - $279.03
Chemical Identifiers
| CAS | 7778-54-3 |
|---|---|
| Molecular Formula | CaCl2O2 |
| Molecular Weight (g/mol) | 142.98 |
| MDL Number | MFCD00010900 |
| InChI Key | ZKQDCIXGCQPQNV-UHFFFAOYSA-N |
| Synonym | calcium hypochlorite, calcium hypochloride, bleaching powder, chlorinated lime, hypochlorous acid, calcium salt, caporit, losantin, pittchlor, pittcide, pittclor |
| PubChem CID | 24504 |
| IUPAC Name | calcium;dihypochlorite |
| SMILES | [O-]Cl.[O-]Cl.[Ca+2] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC300341000
|
Thermo Scientific Chemicals
300341000 |
100 g | Plastic bottle |
Each for $71.93
|
|
||||
|
AC300340010
|
Thermo Scientific Chemicals
300340010 |
1 kg | Plastic bottle |
Each for $279.03
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
| 7778-54-3 | |
| 142.98 | |
| ZKQDCIXGCQPQNV-UHFFFAOYSA-N | |
| 24504 | |
| [O-]Cl.[O-]Cl.[Ca+2] |
| CaCl2O2 | |
| MFCD00010900 | |
| calcium hypochlorite, calcium hypochloride, bleaching powder, chlorinated lime, hypochlorous acid, calcium salt, caporit, losantin, pittchlor, pittcide, pittclor | |
| calcium;dihypochlorite |
Specifications
| 100°C | |
| Green or White | |
| Granular Solid or Granules | |
| CaCl2O2 | |
| MFCD00010900 | |
| 15,1676 | |
| Solubility in water: 200g/L (20°C) (decomposes). Other solubilities: decomposes in alcohol | |
| [O-]Cl.[O-]Cl.[Ca+2] | |
| 142.98 | |
| 142.98 | |
| Technical | |
| Calcium hypochlorite, tech. |
| 7778-54-3 | |
| ∼11.6 (1% aq. soln.) | |
| 100 g | |
| Ca(OCl)2 | |
| 11,107 | |
| calcium hypochlorite, calcium hypochloride, bleaching powder, chlorinated lime, hypochlorous acid, calcium salt, caporit, losantin, pittchlor, pittcide, pittclor | |
| ZKQDCIXGCQPQNV-UHFFFAOYSA-N | |
| calcium;dihypochlorite | |
| 24504 | |
| ≥65% (available Cl) | |
| Plastic bottle |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
May intensify fire; oxidizer.
Contact with acids liberates toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call
GHS Signal Word: Danger
EINECSNumber : 231-908-7
RTECSNumber : NH3485000
TSCA : TSCA
RUO – Research Use Only