missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium hypochlorite, tech., granular, 65% available chlorine
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
| Melting Point | 100°C |
| Color | Green or White |
| pH | ∼11.6 (1% aq. soln.) |
| Physical Form | Granular Solid or Granules |
| Quantity | 1 kg |
| Linear Formula | Ca(OCl)2 |
| Fieser | 11,107 |
| Merck Index | 15,1676 |
| Solubility Information | Solubility in water: 200g/L (20°C) (decomposes). Other solubilities: decomposes in alcohol |
| Formula Weight | 142.98 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes severe skin burns and eye damage. Very toxic to aquatic life. May intensify fire; oxidizer. Contact with acids liberates toxic gas. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call |
| EINECSNumber | 231-908-7 |
| RTECSNumber | NH3485000 |
| TSCA | TSCA |
RUO – Research Use Only