missing translation for 'onlineSavingsMsg'
Learn More

Calcium glycerophosphate hydrate, 97%

CAS: 28917-82-0 | C3H7CaO6P | 210.14 g/mol

$128.70 - $376.01

Chemical Identifiers

CAS 28917-82-0
Molecular Formula C3H7CaO6P
Molecular Weight (g/mol) 210.14
MDL Number MFCD00042643
InChI Key IWIRHXNCFWGFJE-UHFFFAOYNA-L
Synonym calcium glycerophosphate hydrate, calcii glycerophosphas, c3h7o6p.ca.h2o, glycerophosphoric acid calcium salt hydrate, calcium 2,3-dihydroxypropyl phosphate hydrate, calcium dl-glycerol 3-phosphate hydrate
PubChem CID 71311563
IUPAC Name calcium 3-(phosphonatooxy)propane-1,2-diol
SMILES [Ca++].OCC(O)COP([O-])([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1626930
View Documents
Thermo Scientific Chemicals
A1626930
250 g
Each for $128.70
Only null left
 
AAA162690B
View Documents
Thermo Scientific Chemicals
A162690B
1000 g
Each for $376.01
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

28917-82-0
210.14
IWIRHXNCFWGFJE-UHFFFAOYNA-L
71311563
[Ca++].OCC(O)COP([O-])([O-])=O
C3H7CaO6P
MFCD00042643
calcium glycerophosphate hydrate, calcii glycerophosphas, c3h7o6p.ca.h2o, glycerophosphoric acid calcium salt hydrate, calcium 2,3-dihydroxypropyl phosphate hydrate, calcium dl-glycerol 3-phosphate hydrate
calcium 3-(phosphonatooxy)propane-1,2-diol

Specifications

28917-82-0
C3H7CaO6P
14,1670
IWIRHXNCFWGFJE-UHFFFAOYNA-L
calcium 3-(phosphonatooxy)propane-1,2-diol
71311563
97%
250 g
MFCD00042643
calcium glycerophosphate hydrate, calcii glycerophosphas, c3h7o6p.ca.h2o, glycerophosphoric acid calcium salt hydrate, calcium 2,3-dihydroxypropyl phosphate hydrate, calcium dl-glycerol 3-phosphate hydrate
[Ca++].OCC(O)COP([O-])([O-])=O
210.14
210.15 (Anhydrous)
Calcium glycerophosphate hydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 249-312-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only