CAS RN 28917-82-0
CAS RN 28917-82-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Calcium glycerophosphate hydrate, 97%
CAS: 28917-82-0 Molecular Formula: C3H7CaO6P Molecular Weight (g/mol): 210.14 MDL Number: MFCD00042643 InChI Key: IWIRHXNCFWGFJE-UHFFFAOYNA-L Synonym: calcium glycerophosphate hydrate,calcii glycerophosphas,c3h7o6p.ca.h2o,glycerophosphoric acid calcium salt hydrate,calcium 2,3-dihydroxypropyl phosphate hydrate,calcium dl-glycerol 3-phosphate hydrate PubChem CID: 71311563 IUPAC Name: calcium 3-(phosphonatooxy)propane-1,2-diol SMILES: [Ca++].OCC(O)COP([O-])([O-])=O