missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Glycerophosphate, FCC, 98-100.5%, Spectrum™ Chemical
CA147, 27214-00-2, C3H7CaO6P
Supplier: Spectrum Chemical Mfg Cor CA14712KG
Description
Spectrum™ Chemical Calcium Glycerophosphate, FCC is used to neutralize acidic foods when consumed. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 27214-00-2 | |
| C3H7CaO6P | |
| UHHRFSOMMCWGSO-UHFFFAOYSA-L | |
| calcium 2-(phosphonatooxy)propane-1,3-diol | |
| 98 to 100.5% | |
| 12% |
| 100% | |
| 12 kg | |
| [Ca++].OCC(CO)OP([O-])([O-])=O | |
| 210.14 | |
| FCC | |
| Poly Pail |
Chemical Identifiers
| 27214-00-2 | |
| 210.14 | |
| calcium 2-(phosphonatooxy)propane-1,3-diol |
| C3H7CaO6P | |
| UHHRFSOMMCWGSO-UHFFFAOYSA-L | |
| [Ca++].OCC(CO)OP([O-])([O-])=O |