missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Gluconate, Monohydrate, USP, 98.5-102%, Spectrum™ Chemical
CA146, 66905-23-5, C12H22CaO14·H2O
$4109.47 - $4109.47
Chemical Identifiers
| CAS | 66905-23-5 |
|---|---|
| Molecular Formula | C12H24CaO15 |
| Molecular Weight (g/mol) | 448.39 |
| InChI Key | XLNFVCRGJZBQGX-UHFFFAOYNA-L |
| IUPAC Name | calcium bis(2,3,4,5,6-pentahydroxyhexanoate) hydrate |
| SMILES | O.[Ca++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O |
Description
Spectrum™ Chemical Calcium Gluconate, Monohydrate, USP is used as a nutrient. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 66905-23-5 | |
| 448.39 | |
| calcium bis(2,3,4,5,6-pentahydroxyhexanoate) hydrate |
| C12H24CaO15 | |
| XLNFVCRGJZBQGX-UHFFFAOYNA-L | |
| O.[Ca++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O |
Specifications
| 66905-23-5 | |
| 12 kg | |
| XLNFVCRGJZBQGX-UHFFFAOYNA-L | |
| calcium bis(2,3,4,5,6-pentahydroxyhexanoate) hydrate | |
| 98.5 to 102% | |
| 2% |
| 100% | |
| C12H24CaO15 | |
| O.[Ca++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O | |
| 448.39 | |
| USP | |
| Poly Pail |