missing translation for 'onlineSavingsMsg'
Learn More

Calcium Gluconate, Monohydrate, USP, 98.5-102%, Spectrum™ Chemical

Catalog No. 18601307
Change view
Click to view available options
Quantity:
12 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601307 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18601307 Supplier Spectrum Chemical Mfg Cor Supplier No. CA14612KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CA146, 66905-23-5, C12H22CaO14·H2O

Spectrum™ Chemical Calcium Gluconate, Monohydrate, USP is used as a nutrient. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 66905-23-5
Molecular Formula C12H24CaO15
Molecular Weight (g/mol) 448.39
InChI Key XLNFVCRGJZBQGX-UHFFFAOYNA-L
IUPAC Name calcium bis(2,3,4,5,6-pentahydroxyhexanoate) hydrate
SMILES O.[Ca++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O

Specifications

CAS Max % 100%
Physical Form Crystalline Powder or Needles
Quantity 12 kg
Percent Purity 98.5 to 102%
Grade USP
Loss on Drying 2%
Packaging Poly Bottle
Chemical Name or Material Calcium Gluconate, Monohydrate