missing translation for 'onlineSavingsMsg'
Learn More

Calcium Folinate Hydrate 98.0+%, TCI America™

Supplier:  TCI America C22355G

 View more versions of this product

Catalog No. C22355G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Calcium Folinate Hydrate
Yellow
MFCD00006704
calcium folinate
[Ca++].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)N2C=O)N1
511.51
511.51
Crystalline Powder
1492-18-8
C20H21CaN7O7
5 g
KVUAALJSMIVURS-AEQORCIRNA-L
calcium (2S)-2-[(4-{[(2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioate
131675590
≥98.0% (HPLC,T)

Chemical Identifiers

1492-18-8
511.51
KVUAALJSMIVURS-AEQORCIRNA-L
131675590
[Ca++].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)N2C=O)N1
C20H21CaN7O7
MFCD00006704
calcium folinate
calcium (2S)-2-[(4-{[(2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : MA0600500

TSCA : No

SDS
Documents

Documents

Promotions

Promotions