missing translation for 'onlineSavingsMsg'
Learn More

Calcium Fluoride, Powder, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C110512KG

 View more versions of this product

Catalog No. 18609478


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7789-75-5
12 kg
Reagent
Poly Pail
1
97%
0.01

Chemical Identifiers

7789-75-5
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.