missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Fluoride, Powder, 97%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 125 g |
| Percent Purity | 97% |
| Grade | Reagent |
| Loss on Drying | 0.01 |
| Packaging | Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.