missing translation for 'onlineSavingsMsg'
Learn More

Calcium DL-Glycerate Hydrate 98.0+%, TCI America™

Catalog No. G023210G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G023210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G023210G Supplier TCI America Supplier No. G023210G

Chemical Identifiers

CAS 67525-74-0
Molecular Formula C3H5O4
Molecular Weight (g/mol) 105.07
MDL Number MFCD00149289,MFCD00149288,MFCD00149289
InChI Key RBNPOMFGQQGHHO-UWTATZPHSA-M
Synonym calciumdl-glycerate
PubChem CID 131883203
IUPAC Name (2R)-2,3-dihydroxypropanoate
SMILES OC[C@@H](O)C([O-])=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 10 g
Formula Weight 250.22
Percent Purity ≥98.0% (T)
Chemical Name or Material Calcium DL-Glycerate Hydrate