accessibility menu, dialog, popup

UserName

OH(R)OHOO

CAS RN 67525-74-0

IUPAC Name: (2R)-2,3-dihydroxypropanoate
Synonyms: D-glycerate
Molecular Weight (g/mol): 105.07
Molecular Formula: C3H5O4
InChi Key: RBNPOMFGQQGHHO-UWTATZPHSA-M
SMILES: OC[C@@H](O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)