missing translation for 'onlineSavingsMsg'
Learn More

Calcium DL-glycerate dihydrate

Catalog No. AAL0821403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0821403 1 g
AAL0821406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0821403 Supplier Thermo Scientific Chemicals Supplier No. L0821403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 67525-74-0 | C3H5O4 | 105.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in cold and hot water.

Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 67525-74-0
Molecular Formula C3H5O4
Molecular Weight (g/mol) 105.07
MDL Number MFCD00149289,MFCD00149288,MFCD00149289
InChI Key RBNPOMFGQQGHHO-UWTATZPHSA-M
Synonym calciumdl-glycerate
PubChem CID 131883203
IUPAC Name calcium;2,3-dihydroxypropanoic acid;dihydrate
SMILES OC[C@@H](O)C([O-])=O

Specifications

Melting Point ∽140°C (decomposition)
Quantity 1 g
Beilstein 4614104
Merck Index 14,4483
Solubility Information Soluble in cold and hot water.
Formula Weight 286.26 (250.23 Anhydrous)
Chemical Name or Material Calcium DL-glycerate dihydrate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 282-247-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only