missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Disodium EDTA, FCC, 97-102%, Spectrum™ Chemical
E1194, 23411-34-9, C10H12CaN2Na2O8·xH2O
$431.63 - $10998.57
Chemical Identifiers
| CAS | 23411-34-9 |
|---|---|
| Molecular Formula | C10H16CaN2Na2O9 |
| Molecular Weight (g/mol) | 394.30 |
| InChI Key | WJEKUTCBXSHIOX-UHFFFAOYSA-J |
| SMILES | O.[Na+].[Na+].[Ca+4].[O-]C(=O)C[NH](CC[NH](CC([O-])=O)CC([O-])=O)CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18601291
|
Spectrum Chemical Mfg Cor
E1194125GM |
125 g | Poly Bottle |
Each for $431.63
|
|
||||
|
18601292
|
Spectrum Chemical Mfg Cor
E1194500GM |
500 g | Poly Bottle |
Each for $1,281.60
|
|
||||
|
18601293
|
Spectrum Chemical Mfg Cor
E119425KG |
2.5 kg | Poly Pail |
Each for $3,335.90
|
|
||||
|
18601294
|
Spectrum Chemical Mfg Cor
E119445KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Calcium Disodium EDTA, FCC is used as a flavor and color preservative. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 23411-34-9 | |
| 394.30 | |
| O.[Na+].[Na+].[Ca+4].[O-]C(=O)C[NH](CC[NH](CC([O-])=O)CC([O-])=O)CC([O-])=O |
| C10H16CaN2Na2O9 | |
| WJEKUTCBXSHIOX-UHFFFAOYSA-J |
Specifications
| 23411-34-9 | |
| 6.5-7.5 | |
| C10H16CaN2Na2O9 | |
| O.[Na+].[Na+].[Ca+4].[O-]C(=O)C[NH](CC[NH](CC([O-])=O)CC([O-])=O)CC([O-])=O | |
| 97 to 102% | |
| Poly Bottle |
| 100% | |
| 125 g | |
| WJEKUTCBXSHIOX-UHFFFAOYSA-J | |
| 394.30 | |
| FCC |