missing translation for 'onlineSavingsMsg'
Learn More

Medisca Calcium Citrate, USP

Catalog No. 14186113
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186113 2.5 kg
14186161 500 g
14186062 100 g
14186064 1 kg
14186234 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14186113 Supplier Medisca Supplier No. 108001*

Calcium Citrate, USP is a white, odorless, crystalline powder that is slightly hygroscopic.

Calcium Citrate, USP is a white, odorless, crystalline powder that is slightly hygroscopic.

Alternative name(s): Tricalcium citrate

Specifications

Chemical Name or Material Calcium Citrate, USP
Packaging Pail, 8 Liter, White, HDPE, Square
CAS 5785-44-4
Type Drug Standard
Molecular Weight (g/mol) 570.49
Quantity 2.5 kg
Percent Purity 98 to 102 %
Grade USP