Learn More
Calcium Citrate, Tetrahydrate, Powder, FCC, 97.5-100.5%, Spectrum™ Chemical
CA142, 5785-44-4, C12H10Ca3O14·4H2O
Supplier:  Spectrum Chemical Mfg Cor CA14225KG
 View more versions of this product
Catalog No. 18601284
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Calcium Citrate, Tetrahydrate, Powder, FCC is an odorless white powder and the calcium salt of citric acid and is commonly used as a preservative and flavorant food additive. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
5785-44-4
10.0-14%
C12H18Ca3O18
LNIZKKFWMDARJV-UHFFFAOYSA-H
tricalcium bis(2-hydroxypropane-1,2,3-tricarboxylate) tetrahydrate
97.5-100.5%
100%
Poly Pail
2.5 kg
O.O.O.O.[Ca++].[Ca++].[Ca++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
570.49
FCC
Chemical Identifiers
5785-44-4
570.49
tricalcium bis(2-hydroxypropane-1,2,3-tricarboxylate) tetrahydrate
C12H18Ca3O18
LNIZKKFWMDARJV-UHFFFAOYSA-H
O.O.O.O.[Ca++].[Ca++].[Ca++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Product Suggestions
Product Suggestions