missing translation for 'onlineSavingsMsg'
Learn More

Calcium Citrate, Tetrahydrate, Powder, FCC, 97.5-100.5%, Spectrum™ Chemical

Catalog No. 18601283
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
56.7 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601283 500 g Poly Bottle
18601284 2.5 kg Poly Pail
18601285 12 kg Poly Pail
18601286 56.7 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601283 Supplier Spectrum Chemical Mfg Cor Supplier No. CA142500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

CA142, 5785-44-4, C12H10Ca3O14·4H2O

Spectrum™ Chemical Calcium Citrate, Tetrahydrate, Powder, FCC is an odorless white powder and the calcium salt of citric acid and is commonly used as a preservative and flavorant food additive. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5785-44-4
Molecular Formula C12H18Ca3O18
Molecular Weight (g/mol) 570.49
InChI Key LNIZKKFWMDARJV-UHFFFAOYSA-H
IUPAC Name tricalcium bis(2-hydroxypropane-1,2,3-tricarboxylate) tetrahydrate
SMILES O.O.O.O.[Ca++].[Ca++].[Ca++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 100%
Loss on Drying 10.0-14%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 97.5-100.5%
Grade FCC