missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, ultra dry, 99.9% (metals basis)

Catalog No. AA3568504
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3568504 2 g
AA3568509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3568504 Supplier Thermo Scientific Chemicals Supplier No. 03568504

CAS: 10043-52-4 | CaCl2 | 110.98 g/mol

Calcium chloride is used as a drying and dehydrating agent for organic liquids and gases and for solids in desiccators. Common applications include brine for refrigeration plants and ice and dust control on roads. Use of calcium chloride and urea has been reported to significantly increase average fruit weight and ascorbic acid content in pomegranates. The strawberry fruit storability can also be improved by edible coating of calcium chloride. It is used in drying tubes in research laboratories, salt/chemical based dehumidifiers, fire-extinguishers, plastics, ceramic slipware, as flux in the Davy process for the manufacture of sodium and to increase water hardness in swimming pool. It provides inhibition of swelling clays in the water phase of invert emulsion drilling fluids. CaCl2-Pybox (pyridine flanked by two oxazoline groups) has been reported to be an efficient chiral catalyst for asymmetric 1,4-addition reactions of 1,3-dicarbonyl compounds with nitroalkenes, affording gamma-nitro carbonyl compounds in high enantioselectivities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Boiling Point >1600°C
Melting Point 782°C
Physical Form Beads
Quantity 2 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Freely soluble in water,ethanol,methanol,and acetic acid. Insoluble in liquid ammonia,dimethyl sulfoxide and ethyl acetate.
IUPAC Name calcium dichloride
Formula Weight 110.99
Sensitivity Hygroscopic
Density 2.15
Chemical Name or Material Calcium chloride
Show More Show Less
Hazard Category H319-H500
Precautionary Statement P264b-P280i-P305+P351+P338-P312-P501c
EINECSNumber 233-140-8
RTECSNumber EV9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only