missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, Fused, Granulated (0.5 to 2mm), For Elementary Analysis, MilliporeSigma™
$987.46 - $2464.73
Chemical Identifiers
| CAS | 10043-52-4 |
|---|---|
| Molecular Formula | CaCl2 |
| Molecular Weight (g/mol) | 110.98 |
| InChI Key | UXVMQQNJUSDDNG-UHFFFAOYSA-L |
| Synonym | calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal |
| PubChem CID | 5284359 |
| ChEBI | CHEBI:3312 |
| IUPAC Name | calcium dichloride |
| SMILES | [Cl-].[Cl-].[Ca++] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
M1020830250
|
Technidata
1020830250 |
250 g | Plastic bottle |
Each for $987.46
|
|
||||
|
M1020831000
|
Technidata
1020831000 |
1 kg | Plastic bottle |
Each for $2,464.73
|
|
||||
Description
For elementary analysisChemical Identifiers
| 10043-52-4 | |
| 110.98 | |
| calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
| CHEBI:3312 | |
| [Cl-].[Cl-].[Ca++] |
| CaCl2 | |
| UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
| 5284359 | |
| calcium dichloride |
Specifications
| 10043-52-4 | |
| 250 g | |
| calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
| [Cl-].[Cl-].[Ca++] | |
| 110.98 | |
| CHEBI:3312 | |
| Odorless | |
| White solid |
| Solid | |
| CaCl2 | |
| UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
| calcium dichloride | |
| 5284359 | |
| 110.98 | |
| Plastic bottle | |
| Calcium Chloride |
Safety and Handling
H319: Causes serious eye irritation.
EINECSNumber : 233-140-8