missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride, Fused, Granulated (0.5 to 2mm), For Elementary Analysis, MilliporeSigma™

Catalog No. M1020831000
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1020831000 1 kg Plastic bottle
M1020830250 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1020831000 Supplier Technidata Supplier No. 1.02083.1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For elementary analysis

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Physical Form Solid
Quantity 1 kg
Formula Weight 110.98
Odor Odorless
Packaging Plastic bottle
Appearance White solid
Chemical Name or Material Calcium Chloride
Hazard Category H319: Causes serious eye irritation.
EINECSNumber 233-140-8