missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, For BOD, Certified, 2.75% (w/v) ±0.05%, LabChem™
Supplier: LabChem LC127301
Specifications
| Calcium Chloride | |
| For BOD | |
| 96.36,3.64 | |
| Soluble in water | |
| 2.75% ±0.05% (w/v) | |
| Passes Test | |
| Traceable to NIST | |
| 500 mL |
| Colorless | |
| 10035-04-8,7732-18-5 | |
| CaCl2·2H2O | |
| 147.02 | |
| Certified | |
| Poly Bottle | |
| Hydrogen chloride | |
| Liquid |
Chemical Identifiers
| 10035-04-8 |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Refrigerate