missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Dihydrate (White Crystals to Powder), Fisher BioReagents™

Catalog No. p-24969
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP510100 100 g Poly Bottle
BP510250 250 g Poly Bottle
BP510500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP510100 Supplier Fisher BioReagents Supplier No. FLBP510 100
Only null left

Calcium Chloride Dihydrate (White Crystals to Powder)

Calcium Chloride is suitable for use in the preparation of buffer solutions.

Chemical Identifiers

CAS 10035-04-8 , 10043-52-4
Molecular Formula Ca Cl2 · 2 H2O

Specifications

Melting Point 175°C
Color Beige-White
pH 4.5 to 6.5
Physical Form Solid
Quantity 100 g
Assay Percent Range 99.0 to 105.0 %
Merck Index 15, 1661
Formula Weight 147.02
Percent Purity 99.0 to 105.0%
Identification Pass Test
Packaging Poly Bottle
Density 0.830g/cm³
Chemical Name or Material Calcium Chloride Dihydrate
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:1
Recommended Storage RT