Learn More
Calcium Chloride Dihydrate (White Crystals to Powder), Fisher BioReagents™
Supplier: Fisher BioReagents FLBP510250
Description
Calcium Chloride Dihydrate (White Crystals to Powder)
Calcium Chloride is suitable for use in the preparation of buffer solutions.Specifications
| Calcium Chloride Dihydrate | |
| Beige-White | |
| 4.5 to 6.5 | |
| 250 g | |
| Ca Cl2 · 2 H2O | |
| 147.02 | |
| Pass Test | |
| 0.830g/cm³ |
| 175°C | |
| 10035-04-8,10043-52-4 | |
| Solid | |
| 99.0 to 105.0 % | |
| 15, 1661 | |
| 99.0 to 105.0% | |
| Poly Bottle |
Chemical Identifiers
| 10035-04-8 | |
| Ca Cl2 · 2 H2O |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:2
Flammability:0
Instability:1
Recommended Storage : RT