missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride (Desiccant, 20 mesh and Finer) ACS Grade, Fisher Chemical™

Catalog No. p-31481
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C77500 500 g Poly Bottle
C77212 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C77500 Supplier Thermo Fisher Scientific Supplier No. FLC77500
Only null left

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Physical Form Solid
Melting Point 782°C
pH 8 to 10
Boiling Point 1600°C
Color Beige
Density 2.15g/mL
Material Calcium Chloride
Quantity 500 g
Identification Pass Test
Assay Percent Range ≥96 %
Mesh Size 20 mesh and finer
Packaging Poly Bottle
Formula Weight 110.99
Percent Purity ≥96%
Grade ACS Grade
Chemical Name or Material Calcium Chloride, Desiccant
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:1